Bis(acetonitrile)palladium(II) dichloride (40.5% Pd)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

10002.03 10002.06
10002.03EA 124 GBP
10002.03 10002.06
Bis(acetonitrile)palladium(II) dichloride (40.5% Pd)
Bis(acetonitrile)palladium(II) dichloride

CAS No.: 14592-56-4 Molecular Formula: PdCl2(CH3CN)2 Formula Weight: 259.41 Physical Form: Powder Odour: Odourless Solubility: Soluble in acetonitrile, acetone, chloroform UN No.: UN3439 MDL No.: MFCD00013122


Applications: Olefin isomerization, preparation of acetals and hemiacetal esters

Formula: C₄H₆Cl₂N₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00013122
CAS Number: 14592-56-4
EINECS: 238-637-3
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR